Gene Transcripts

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
Chromosome Location
Chr. 19: 18001132 - 18014102 on Build GRCh38
UniGene ID
Hs.515249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025604.2 NP_001020775.1 5 971
NM_001286826.1 NP_001273755.1 5 946
NM_015683.1 NP_056498.1 5 946
GenBank mRNA AF131826.1 - 4 478
AF193051.1 - 5 946
AK127940.1 - 5 935
AK128588.1 - 1 1824
AK314657.1 - 5 944
BC022516.1 - 5 944 4817429

View Full Product Details