Gene Transcripts
- Gene Symbol
- CAND1.11
- Entrez Gene ID
- 100130460
- Gene Name
- uncharacterized LOC100130460
- Gene Aliases
- Chromosome Location
- Chr. 11: 10308313 - 10430673 on Build GRCh38
- UniGene ID
- Hs.501890
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103765.1 |
|
4 |
740 |
|
GenBank mRNA |
AI652043.1 |
- |
1 |
277 |
|
BX090685.1 |
- |
3 |
399 |
|
View Full Product Details