Gene Transcripts

Gene Symbol
C7orf72
Entrez Gene ID
100130988
Gene Name
chromosome 7 open reading frame 72
Gene Aliases
Chromosome Location
Chr. 7: 50095861 - 50159256 on Build GRCh38
UniGene ID
Hs.99248
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161834.2 NP_001155306.2 1 98
XM_011515052.1 XP_011513354.1 1 323
XM_011515053.2 XP_011513355.1 1 323

View Full Product Details