Gene Transcripts

Gene Symbol
ARHGEF37
Entrez Gene ID
389337
Gene Name
Rho guanine nucleotide exchange factor 37
Gene Aliases
Chromosome Location
Chr. 5: 149552049 - 149634968 on Build GRCh38
UniGene ID
Hs.256206
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001669.2 NP_001001669.2 12 1739
XM_005268447.4 XP_005268504.1 12 1710
XM_005268448.1 XP_005268505.1 11 1613
XM_006714784.3 XP_006714847.1 13 2078
XM_011537642.2 XP_011535944.1 12 1761
XM_017009460.1 XP_016864949.1 11 1852
XM_017009461.1 XP_016864950.1 10 1502
GenBank mRNA AB199796.1 - 11 1686
AK123597.1 - 4 379
AK300355.1 - 11 1600
AK300559.1 - 12 1838
AK316002.1 - 12 1809
BF510234.1 - 2 41

View Full Product Details