Gene Transcripts

Gene Symbol
CFAP46
Entrez Gene ID
54777
Gene Name
cilia and flagella associated protein 46
Gene Aliases
C10orf123, C10orf124, C10orf92, C10orf93, TTC40, bA288G11.4, bA288G11.5, bB137A17.2, bB137A17.3
Chromosome Location
Chr. 10: 132808392 - 132942585 on Build GRCh38
UniGene ID
Hs.298038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001200049.2 NP_001186978.2 27 3694
GenBank mRNA DN930461.1 - 2 80
DY654537.1 - 1 154

View Full Product Details