Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145046.4 | NP_659483.2 | 1 | 133 | |
GenBank mRNA | AB576176.1 | - | 1 | 70 | |
AK058084.1 | - | 1 | 125 | ||
BC014595.2 | - | 1 | 133 | 4822010 | |
BP370084.1 | - | 1 | 113 | ||
DB459545.1 | - | 1 | 135 |