Gene Transcripts

Gene Symbol
ADRA2C
Entrez Gene ID
152
Gene Name
adrenoceptor alpha 2C
Gene Aliases
ADRA2L2, ADRA2RL2, ADRARL2, ALPHA2CAR
Chromosome Location
Chr. 4: 3766569 - 3768526 on Build GRCh38
UniGene ID
Hs.123022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000683.3 NP_000674.2 1 713
GenBank mRNA BC142625.1 - 1 884
J03853.1 - 1 713

View Full Product Details