Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033583.2 | NP_001028755.2 | 7 | 689 | |
NM_001037171.1 | NP_001032248.1 | 8 | 716 | ||
XM_005274472.2 | XP_005274529.1 | 6 | 588 | ||
GenBank mRNA | AF132950.1 | - | 6 | 591 | |
AK000659.1 | - | 7 | 675 | ||
AK024337.1 | - | 9 | 1781 | ||
AL548456.3 | - | 8 | 716 | ||
BC012573.1 | - | 7 | 660 | ||
BC136671.1 | - | 7 | 640 | 9020661 | |
BC144360.1 | - | 7 | 640 | 9052881 | |
BC144370.1 | - | 7 | 640 | ||
BX648512.1 | - | 8 | 700 |