Gene Transcripts

Gene Symbol
C10orf12
Entrez Gene ID
26148
Gene Name
chromosome 10 open reading frame 12
Gene Aliases
Chromosome Location
Chr. 10: 96981284 - 96985828 on Build GRCh38
UniGene ID
Hs.427927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015652.2 NP_056467.2 1 164
GenBank mRNA AK126886.1 - 1 217
BC024315.1 - 1 164 3903728

View Full Product Details