Gene Transcripts

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
Chromosome Location
Chr. 9: 114334149 - 114398498 on Build GRCh38
UniGene ID
Hs.494895
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317950.1 NP_001304879.1 10 2476
NM_001317952.1 NP_001304881.1 9 2034
NM_030767.5 NP_110394.3 10 2421
XM_005252244.2 XP_005252301.1 12 2593
XM_005252245.1 XP_005252302.1 11 2451
XM_005252247.4 XP_005252304.1 10 2472
XM_005252248.2 XP_005252305.1 10 2589
XM_006717294.1 XP_006717357.1 10 2405
XM_006717295.2 XP_006717358.1 10 2589
XM_011519063.2 XP_011517365.1 9 2084
XM_011519064.2 XP_011517366.1 10 2589
XM_011519065.2 XP_011517367.2 10 2064
XM_011519066.2 XP_011517368.1 10 2589
XM_017015172.1 XP_016870661.1 11 2191
GenBank mRNA AK024431.1 - 8 1369
AK074040.1 - 9 1464
AL833220.1 - 7 898
AY703039.1 - 11 2076
AY703040.1 - 9 2034
AY703041.1 - 11 1790
AY703042.1 - 9 1603
AY703043.1 - 9 2394
AY703044.1 - 8 2088
BC055285.1 - 10 2421 6143155

View Full Product Details