Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052855.3 | NP_443087.1 | 1, 2 | 400 | |
XM_017025296.1 | XP_016880785.1 | 1, 2 | 147 | ||
GenBank mRNA | AK314010.1 | - | 1, 2 | 147 | |
BC012978.2 | - | 1, 2 | 392 | 3834331 | |
CR627413.1 | - | 1, 2 | 172 | ||
EU164539.1 | - | 1, 2 | 392 |