Gene Transcripts

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
Chromosome Location
Chr. 9: 69324572 - 69392461 on Build GRCh38
UniGene ID
Hs.118003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127608.1 NP_001121080.1 6, 7 605
NM_004816.3 NP_004807.3 6, 7 921
XM_005252307.4 XP_005252364.1 6, 7 1482
XM_017015324.1 XP_016870813.1 5, 6 666
XM_017015325.1 XP_016870814.1 6, 7 605
GenBank mRNA AK056152.1 - 4, 5 549
AK126127.1 - 6, 7 921
BC021685.1 - 3, 4 334
BC113683.1 - 6, 7 564 8322735
BC113685.1 - 6, 7 564 8322737
BX641153.1 - 3, 4 5774

View Full Product Details