Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159536.1 | NP_001153008.1 | 2 | 1006 | |
NM_001159537.1 | NP_001153009.1 | 2 | 1065 | ||
NM_138305.3 | NP_612178.2 | 1 | 683 | ||
GenBank mRNA | AF458089.1 | - | 1 | 655 | |
AK122298.1 | - | 2 | 977 | ||
AK202634.1 | - | 1 | 105 | ||
BC057113.1 | - | 2 | 994 | 6835448 | |
BC057316.1 | - | 2 | 895 | 6830961 |