Gene Transcripts
- Gene Symbol
- Bsg
- Entrez Gene ID
- 12215
- Gene Name
- basigin
- Gene Aliases
- AI115436, AI325119, CD147, EMMPRIN, HT-7
- Chromosome Location
- Chr. 10: 79704358 - 79711979 on Build GRCm38
- UniGene ID
- Mm.726
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077184.1 |
NP_001070652.1
|
4, 5 |
612 |
|
NM_009768.2 |
NP_033898.1
|
5, 6 |
960 |
|
GenBank mRNA |
AK002332.1 |
- |
4, 5 |
464 |
|
AK011364.1 |
- |
4, 5 |
463 |
|
AK147225.1 |
- |
4, 5 |
5286 |
|
AK207023.1 |
- |
2, 3 |
211 |
|
AK209680.1 |
- |
2, 3 |
212 |
|
AK215038.1 |
- |
2, 3 |
212 |
|
AK216955.1 |
- |
2, 3 |
212 |
|
AY089967.1 |
- |
5, 6 |
818 |
|
BC010270.1 |
- |
4, 5 |
455 |
3589236 |
D00611.1 |
- |
4, 5 |
462 |
|
S63813.1 |
- |
4, 5 |
462 |
|
Y16256.1 |
- |
4, 5 |
462 |
|
Y16257.1 |
- |
3, 4 |
458 |
|
Y16258.1 |
- |
3, 4 |
404 |
|
View Full Product Details