Gene Transcripts

Gene Symbol
Aox2
Entrez Gene ID
213043
Gene Name
aldehyde oxidase 2
Gene Aliases
Aoh3, Aox3l1
Chromosome Location
Chr. 1: 58276141 - 58379264 on Build GRCm38
UniGene ID
Mm.414292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008419.2 NP_001008419.1 9 793
GenBank mRNA AY187018.2 - 9 793
AY665589.1 - 9 700
BC150826.1 - 9 720 9087737

View Full Product Details