Gene Transcripts

Gene Symbol
Eif2b2
Entrez Gene ID
217715
Gene Name
eukaryotic translation initiation factor 2B, subunit 2 beta
Gene Aliases
AA409345, C85417, EIF-2Bbeta, EIF2B
Chromosome Location
Chr. 12: 85219475 - 85226628 on Build GRCm38
UniGene ID
Mm.29041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145445.3 NP_663420.1 8 1056
GenBank mRNA AK049731.1 - 8 1057
AK076195.1 - 8 1052
BC003326.1 - 8 1088 3156632

View Full Product Details