Gene Transcripts

Gene Symbol
Bpifb1
Entrez Gene ID
228801
Gene Name
BPI fold containing family B, member 1
Gene Aliases
Lplunc1
Chromosome Location
Chr. 2: 154190818 - 154220343 on Build GRCm38
UniGene ID
Mm.3783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012392.1 NP_001012392.1 14, 15 1431
NM_153418.2 NP_700467.2 14, 15 1394
GenBank mRNA BC060982.1 - 14, 15 1431
BC100436.1 - 14, 15 1426 30298932
U46068.3 - 14, 15 1395

View Full Product Details