Gene Transcripts

Gene Symbol
Bpifb5
Entrez Gene ID
228802
Gene Name
BPI fold containing family B, member 5
Gene Aliases
Lplunc5
Chromosome Location
Chr. 2: 154223742 - 154240902 on Build GRCm38
UniGene ID
Mm.167411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144890.2 NP_659139.2 3, 4 481
GenBank mRNA AK142982.1 - 3, 4 482
BC018465.1 - 3, 4 443 3156626

View Full Product Details