Gene Transcripts

Gene Symbol
Sowahd
Entrez Gene ID
245381
Gene Name
sosondowah ankyrin repeat domain family member D
Gene Aliases
A630014H24Rik, Ankrd58
Chromosome Location
Chr. X: 37048845 - 37050418 on Build GRCm38
UniGene ID
Mm.123697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173779.3 NP_776140.1 1 829
GenBank mRNA AK041485.1 - 1 830
BC107372.1 - 1 827 40056413
BC107373.1 - 1 828 40056414

View Full Product Details