Gene Transcripts

Gene Symbol
Akap17b
Entrez Gene ID
338351
Gene Name
A kinase (PRKA) anchor protein 17B
Gene Aliases
AA407619, AKAP-17B, B230333C21Rik, C230056F04Rik, PRKA17B, Sfrs17b, Srsf17b, Talia
Chromosome Location
Chr. X: 36608173 - 36645471 on Build GRCm38
UniGene ID
Mm.116704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081956.1 NP_001075425.1 3 949
GenBank mRNA AK046007.1 - 3 931
AK054091.1 - 3 952

View Full Product Details