Gene Transcripts

Gene Symbol
Asb12
Entrez Gene ID
70392
Gene Name
ankyrin repeat and SOCS box-containing 12
Gene Aliases
2310036C05Rik, Asb-12
Chromosome Location
Chr. X: 95470199 - 95557993 on Build GRCm38
UniGene ID
Mm.79122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080858.3 NP_543134.1 2 689
GenBank mRNA AF403040.1 - 2 690
AK009644.1 - 2 690
BC107257.2 - 2 654 40052462

View Full Product Details