Gene Transcripts

Gene Symbol
Ankrd53
Entrez Gene ID
75305
Gene Name
ankyrin repeat domain 53
Gene Aliases
4930564N15Rik
Chromosome Location
Chr. 6: 83762646 - 83768326 on Build GRCm38
UniGene ID
Mm.391478
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029245.3 NP_083521.2 6 1411
GenBank mRNA AK016222.1 - 5 1207
AK133099.1 - 6 1413

View Full Product Details