Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161667.1 | NP_001155139.1 | 3 | 379 | |
NM_053115.2 | NP_444345.2 | 3 | 504 | ||
GenBank mRNA | AJ238492.1 | - | 3 | 421 | |
AK144243.1 | - | 3 | 373 | ||
BC021339.1 | - | 3 | 383 | 5052291 |