Gene Transcripts
- Gene Symbol
- Olfr1135
- Entrez Gene ID
- 258654
- Gene Name
- olfactory receptor 1135
- Gene Aliases
- MOR177-2
- Chromosome Location
- Chr. 2: 87671433 - 87672365 on Build GRCm38
- UniGene ID
- Mm.377617
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146660.2 |
NP_666871.2
|
1 |
249 |
|
View Full Product Details