Gene Transcripts

Gene Symbol
Batf2
Entrez Gene ID
74481
Gene Name
basic leucine zipper transcription factor, ATF-like 2
Gene Aliases
4933430F08Rik
Chromosome Location
Chr. 19: 6164392 - 6172476 on Build GRCm38
UniGene ID
Mm.159219
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028967.1 NP_083243.1 3 482
GenBank mRNA AK016990.1 - 698
AK150133.1 - 3 502
AK171828.1 - 3 499
BC024521.1 - 3 482 4984403

View Full Product Details