Gene Transcripts

Gene Symbol
Mrps26
Entrez Gene ID
99045
Gene Name
mitochondrial ribosomal protein S26
Gene Aliases
AI648866, GI008, MRP-S13, MRP-S26, Rpms13
Chromosome Location
Chr. 2: 130563757 - 130565394 on Build GRCm38
UniGene ID
Mm.30057
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207207.1 NP_997090.1 4 539
GenBank mRNA AK131626.1 - 3 629
AK169837.1 - 4 552
AK213019.1 - 3 178
BC048517.1 - 4 539 6706680

View Full Product Details