Gene Transcripts

Gene Symbol
Bmp2k
Entrez Gene ID
498333
Gene Name
BMP-2 inducible kinase
Gene Aliases
Chromosome Location
Chr. 14: 14126086 - 14226823 on Build Rnor_6.0
UniGene ID
Rn.234368
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770127.1 XP_008768349.1 9 1563
GenBank mRNA BC103651.1 - 2 202

View Full Product Details