Gene Transcripts

Gene Symbol
Acsbg2
Entrez Gene ID
301120
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
Chromosome Location
Chr. 9: 10172331 - 10203550 on Build Rnor_6.0
UniGene ID
Rn.109833
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080096.1 NP_001073565.1 11 1687
XM_006244311.2 XP_006244373.1 11 2196
XM_006244312.2 XP_006244374.1 11 2198
GenBank mRNA BC129110.1 - 11 1687

View Full Product Details