Gene Transcripts

Gene Symbol
Bmi1
Entrez Gene ID
307151
Gene Name
Bmi1 polycomb ring finger oncogene
Gene Aliases
Pcgf4
Chromosome Location
Chr. 17: 85361598 - 85370283 on Build Rnor_6.0
UniGene ID
Rn.220522
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107368.2 NP_001100838.1 10 788
XM_008771871.1 XP_008770093.1 10 3673
GenBank mRNA BC168209.1 - 10 788

View Full Product Details