Gene Transcripts

Gene Symbol
Agmo
Entrez Gene ID
362732
Gene Name
alkylglycerol monooxygenase
Gene Aliases
RGD1312038, Tmem195
Chromosome Location
Chr. 6: 56846859 - 57193918 on Build Rnor_6.0
UniGene ID
Rn.152690
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135899.1 NP_001129371.1 5 688
GenBank mRNA BC127545.1 - 5 688
FQ211046.1 - 5 744
FQ219104.1 - 5 732

View Full Product Details