Gene Transcripts

Gene Symbol
Ccdc64
Entrez Gene ID
304537
Gene Name
coiled-coil domain containing 64
Gene Aliases
RGD1310292
Chromosome Location
Chr. 12: 46615584 - 46703288 on Build Rnor_6.0
UniGene ID
Rn.166161
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191667.1 NP_001178596.1 4 828
XM_008769234.1 XP_008767456.1 4 893

View Full Product Details