Gene Transcripts

Gene Symbol
Bai1
Entrez Gene ID
362931
Gene Name
brain-specific angiogenesis inhibitor 1
Gene Aliases
Chromosome Location
Chr. 7: 115766484 - 115828514 on Build Rnor_6.0
UniGene ID
Rn.103502
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170597.2 NP_001164068.2 6 1700

View Full Product Details