Gene Transcripts

Gene Symbol
C1qtnf2
Entrez Gene ID
497886
Gene Name
C1q and tumor necrosis factor related protein 2
Gene Aliases
Ctrp2, RGD1561041
Chromosome Location
Chr. 10: 29067102 - 29084035 on Build Rnor_6.0
UniGene ID
Rn.107489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191918.1 NP_001178847.1 4 876
XM_006246141.2 XP_006246203.1 4 963

View Full Product Details