Gene Transcripts

Gene Symbol
Ccdc3
Entrez Gene ID
498795
Gene Name
coiled-coil domain containing 3
Gene Aliases
RGD1559473
Chromosome Location
Chr. 17: 76991722 - 77093522 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_574081.6 XP_574081.4 2, 3 753

View Full Product Details