Gene Transcripts

Gene Symbol
Sowahc
Entrez Gene ID
503306
Gene Name
sosondowah ankyrin repeat domain family member C
Gene Aliases
Ankrd57
Chromosome Location
Chr. 20: 28815039 - 28819505 on Build Rnor_6.0
UniGene ID
Rn.55986
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109364.1 NP_001102834.1 1 241

View Full Product Details