Gene Transcripts

Gene Symbol
Sost
Entrez Gene ID
80722
Gene Name
sclerostin
Gene Aliases
Chromosome Location
Chr. 10: 89897087 - 89900131 on Build Rnor_6.0
UniGene ID
Rn.95369
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030584.1 NP_085073.1 2 286
GenBank mRNA AF326741.1 - 2 286

View Full Product Details