Gene Transcripts

Gene Symbol
CASC2
Entrez Gene ID
255082
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
Gene Aliases
C10orf5
Chromosome Location
Chr. 10: 118046821 - 118210153 on Build GRCh38
UniGene ID
Hs.89387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026939.1 1 96
NR_026940.1 1 96
NR_026941.1 1 96
GenBank mRNA BC036393.1 - 1 98

View Full Product Details