Gene Transcripts

Gene Symbol
C3P1
Entrez Gene ID
388503
Gene Name
complement component 3 precursor pseudogene
Gene Aliases
CPLP
Chromosome Location
Chr. 19: 10041356 - 10074138 on Build GRCh38
UniGene ID
Hs.655363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027300.2 18 2286
GenBank mRNA AK131489.1 - 16 2051

View Full Product Details