Gene Transcripts
- Gene Symbol
- LINC00575
- Entrez Gene ID
- 439934
- Gene Name
- long intergenic non-protein coding RNA 575
- Gene Aliases
- C4orf11
- Chromosome Location
- Chr. 4: 82613113 - 82621437 on Build GRCh38
- UniGene ID
- Hs.457189
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000426551.5 |
- |
2 |
386 |
|
Chr. 4 - 82613113 - 82621437 [-] |
GENCODE |
ENST00000515811.1 |
- |
2 |
324 |
|
Chr. 4 - 82613113 - 82621437 [-] |
View Full Product Details