Gene Transcripts

Gene Symbol
NOP14-AS1
Entrez Gene ID
317648
Gene Name
NOP14 antisense RNA 1
Gene Aliases
C4orf10, RES4-24
Chromosome Location
Chr. 4: 2935546 - 2951078 on Build GRCh38
UniGene ID
Hs.398178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015453.2 1 190
GenBank mRNA AB000464.1 - 1 190
AB000466.1 - 1 190
BC020978.1 - 1 812

View Full Product Details