Gene Transcripts
- Gene Symbol
- ASMTL-AS1
- Entrez Gene ID
- 80161
- Gene Name
- ASMTL antisense RNA 1
- Gene Aliases
- ASMTL-AS, ASMTLAS, CXYorf2, NCRNA00105
- Chromosome Location
- Chr. Y: 1400531 - 1415421 on Build GRCh38
- UniGene ID
- Hs.521856
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026710.1 |
|
2 |
235 |
|
NR_026711.1 |
|
2 |
235 |
|
View Full Product Details