Gene Transcripts

Gene Symbol
LINC00597
Entrez Gene ID
81698
Gene Name
long intergenic non-protein coding RNA 597
Gene Aliases
C15orf5
Chromosome Location
Chr. 15: 77223893 - 77225404 on Build GRCh38
UniGene ID
Hs.656938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026813.1 1 858
GenBank mRNA AF216224.1 - 1 864
AL109679.1 - 1 503
BC069765.1 - 1 794

View Full Product Details