Gene Transcripts
- Gene Symbol
- ANKFY1
- Entrez Gene ID
- 51479
- Gene Name
- ankyrin repeat and FYVE domain containing 1
- Gene Aliases
- ANKHZN, BTBD23, ZFYVE14
- Chromosome Location
- Chr. 17: 4163371 - 4264026 on Build GRCh38
- UniGene ID
- Hs.696087
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091008.1 |
- |
1 |
669 |
|
View Full Product Details