Gene Transcripts
- Gene Symbol
- BCAS3
- Entrez Gene ID
- 54828
- Gene Name
- BCAS3, microtubule associated cell migration factor
- Gene Aliases
- GAOB1, MAAB
- Chromosome Location
- Chr. 17: 60677811 - 61392838 on Build GRCh38
- UniGene ID
- Hs.655028
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125409.1 |
- |
1 |
918 |
|
View Full Product Details