Gene Transcripts
- Gene Symbol
- LOC100505736
- Entrez Gene ID
- 100505736
- Gene Name
- uncharacterized LOC100505736
- Gene Aliases
- Chromosome Location
- Chr. 2: 28148555 - 28206961 on Build GRCh38
- UniGene ID
- Hs.659540
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120504.1 |
|
3 |
937 |
|
GenBank mRNA |
AK057415.1 |
- |
3 |
940 |
|
View Full Product Details