Gene Transcripts
- Gene Symbol
- C6
- Entrez Gene ID
- 729
- Gene Name
- complement component 6
- Gene Aliases
- Chromosome Location
- Chr. 5: 41142116 - 41261486 on Build GRCh38
- UniGene ID
- Hs.481992
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
J05063.1 |
- |
1 |
249 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX502917.1 |
- |
1 |
102 |
|
Chr. 5 - 41159261 - 41159994 [-] |
View Full Product Details