Gene Transcripts
- Gene Symbol
- ARMC8
- Entrez Gene ID
- 25852
- Gene Name
- armadillo repeat containing 8
- Gene Aliases
- GID5, HSPC056, S863-2, VID28
- Chromosome Location
- Chr. 3: 138187248 - 138298389 on Build GRCh38
- UniGene ID
- Hs.744868
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128702.1 |
- |
1 |
2290 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN293842.1 |
- |
1 |
48 |
|
Chr. 3 - 138261036 - 138261651 [+] |
View Full Product Details