Gene Transcripts
- Gene Symbol
- 2310016D03Rik
- Entrez Gene ID
- 69566
- Gene Name
- RIKEN cDNA 2310016D03 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 30410559 - 30467358 on Build GRCm38
- UniGene ID
- Mm.396405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045491.1 |
|
3 |
1346 |
|
GenBank mRNA |
AK009380.1 |
- |
3 |
1347 |
|
View Full Product Details