Gene Transcripts
- Gene Symbol
- Gm20609
- Entrez Gene ID
- 102637684
- Gene Name
- predicted gene 20609
- Gene Aliases
- Chromosome Location
- Chr. 10: 71310745 - 71313977 on Build GRCm38
- UniGene ID
- Mm.338906
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK158974.1 |
- |
4 |
1221 |
|
View Full Product Details