Gene Transcripts
- Gene Symbol
- Ammecr1l
- Entrez Gene ID
- 225339
- Gene Name
- AMME chromosomal region gene 1-like
- Gene Aliases
- 5430429D03Rik, AU022236, AU040755, AW111353, E230022H04Rik
- Chromosome Location
- Chr. 18: 31759475 - 31784083 on Build GRCm38
- UniGene ID
- Mm.290804
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017359.1 |
- |
1 |
1068 |
|
AK049081.1 |
- |
1 |
1135 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI251559.1 |
- |
1 |
472 |
|
Chr. 18 - 31783181 - 31783836 [+] |
View Full Product Details